siRNA Details
| Gene Symbol: Irgq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153134.3 | NP_694774.3 | 3 | 1294 | ||
| GenBank mRNA | AF322649.1 | - | 2 | 1083 | ||
| AK051117.1 | - | 3 | 1204 | |||
| AK210871.1 | - | 1 | 223 | |||
| BC056347.1 | - | 3 | 1253 | 5705226 | ||
| BC080294.1 | - | 3 | 1232 | 6856174 | ||

Irgq
Genomic Map