siRNA Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252478.1 | NP_001239407.1 | 8 | 488 | ||
| NM_144798.6 | NP_659047.2 | 8 | 473 | |||
| GenBank mRNA | AF233346.1 | - | 7 | 425 | ||
| AF395840.1 | - | 7 | 415 | |||
| AK082907.1 | - | 8 | 450 | |||
| AK150342.1 | - | 8 | 449 | |||
| AK151120.1 | - | 8 | 449 | |||
| AK160903.1 | - | 8 | 477 | |||
| BC066162.1 | - | 8 | 440 | 30471027 | ||
| BC068169.1 | - | 8 | 462 | 30542277 | ||

Slc30a6
Genomic Map