siRNA Details
| Gene Symbol: Gm4763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177593.1 | NP_808261.1 | 1, 2 | 100 | ||
| GenBank mRNA | BC037749.1 | - | 1, 2 | 100 | 1429110 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4763| Gene Symbol: Gm4763 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177593.1 | NP_808261.1 | 1, 2 | 100 | ||
| GenBank mRNA | BC037749.1 | - | 1, 2 | 100 | 1429110 | |