siRNA Details
| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175436.4 | NP_780645.1 | 3 | 1757 | ||
| GenBank mRNA | AK050839.1 | - | 3 | 1758 | ||
| AK051798.1 | - | 3 | 1755 | |||
| BC052548.1 | - | 2 | 1711 | |||
| BC052795.1 | - | 1 | 857 | |||

Zfp526
Genomic Map