siRNA Details
| Gene Symbol: Gm12169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163357.1 | NP_001156829.1 | 3 | 240 | ||
| GenBank mRNA | BG079892.2 | - | 3 | 225 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12169| Gene Symbol: Gm12169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163357.1 | NP_001156829.1 | 3 | 240 | ||
| GenBank mRNA | BG079892.2 | - | 3 | 225 | ||