siRNA Details
| Gene Symbol: Hus1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153072.2 | NP_694712.1 | 1 | 656 | ||
| GenBank mRNA | AF508546.1 | - | 1 | 569 | ||
| AK133107.1 | - | 1 | 658 | |||
| BC125607.1 | - | 1 | 649 | 40130122 | ||
| BC125609.1 | - | 1 | 649 | 40130124 | ||

Hus1b
Genomic Map