siRNA Details
| Gene Symbol: Pamr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173749.4 | NP_776110.3 | 1, 2 | 250 | ||
| GenBank mRNA | AK031254.1 | - | 1, 2 | 123 | ||
| AK088017.1 | - | 1, 2 | 149 | |||
| BB620582.1 | - | 1, 2 | 117 | |||
| BC031841.1 | - | 1, 2 | 204 | 3675412 | ||
| BC057685.1 | - | 1, 2 | 204 | 3675412 | ||
| BY266233.1 | - | 1, 2 | 200 | |||

Pamr1
Genomic Map