siRNA Details
| Gene Symbol: Unc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153135.3 | NP_694775.1 | 14, 15 | 2478 | ||
| GenBank mRNA | AJ487854.1 | - | 14, 15 | 2479 | ||
| AK220417.1 | - | 4, 5 | 819 | |||
| BC137615.1 | - | 14, 15 | 2269 | 8860634 | ||
| BC144783.1 | - | 12, 13 | 2086 | 9053331 | ||
| BC144784.1 | - | 12, 13 | 2047 | 9053332 | ||
| BC145695.1 | - | 11, 12 | 2053 | 40130998 | ||
| FJ824810.1 | - | 13, 14 | 2357 | |||

Unc5d
Genomic Map