siRNA Details
| Gene Symbol: Zfp947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177596.3 | NP_808264.1 | 5 | 1024 | ||
| GenBank mRNA | AK032784.1 | - | 5 | 1024 | ||
| BC120908.1 | - | 4 | 761 | 40088624 | ||
| BC120909.1 | - | 4 | 761 | 40088625 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp947| Gene Symbol: Zfp947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177596.3 | NP_808264.1 | 5 | 1024 | ||
| GenBank mRNA | AK032784.1 | - | 5 | 1024 | ||
| BC120908.1 | - | 4 | 761 | 40088624 | ||
| BC120909.1 | - | 4 | 761 | 40088625 | ||