siRNA Details
Gene Symbol: D130040H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172491.2 | NP_766079.1 | 6 | 1196 | ||
GenBank mRNA | AK051365.1 | - | 6 | 1196 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D130040H23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172491.2 | NP_766079.1 | 6 | 1196 | ||
GenBank mRNA | AK051365.1 | - | 6 | 1196 |