siRNA Details
| Gene Symbol: D130040H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172491.2 | NP_766079.1 | 6 | 1449 | ||
| GenBank mRNA | AK051365.1 | - | 6 | 1449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D130040H23Rik| Gene Symbol: D130040H23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172491.2 | NP_766079.1 | 6 | 1449 | ||
| GenBank mRNA | AK051365.1 | - | 6 | 1449 | ||