siRNA Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170673.3 | NP_733773.2 | 4 | 339 | ||
| GenBank mRNA | AK042692.1 | - | 4 | 340 | ||
| AK044080.1 | - | 4 | 305 | |||
| AK047431.1 | - | 1 | 1031 | |||
| AK047571.1 | - | 4 | 283 | |||
| AK137422.1 | - | 4 | 340 | |||
| BB639866.1 | - | 4 | 340 | |||
| BC115728.1 | - | 4 | 289 | 40039295 | ||

Cpne9
Genomic Map