siRNA Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170673.3 | NP_733773.2 | 10, 11 | 828 | ||
| GenBank mRNA | AK042692.1 | - | 10, 11 | 829 | ||
| AK044080.1 | - | 10, 11 | 794 | |||
| AK047571.1 | - | 10, 11 | 772 | |||
| AK137422.1 | - | 10, 11 | 829 | |||
| AK220550.1 | - | 3, 4 | 1765 | |||
| BC115728.1 | - | 10, 11 | 778 | 40039295 | ||

Cpne9
Genomic Map