siRNA Details
| Gene Symbol: Cln5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033242.1 | NP_001028414.1 | 2, 3 | 415 | ||
| GenBank mRNA | AK032293.1 | - | 2, 3 | 230 | ||
| AK043247.1 | - | 2, 3 | 339 | |||
| AK085741.1 | - | 2, 3 | 416 | |||
| AK144635.1 | - | 2, 3 | 306 | |||
| AK207183.1 | - | 1, 2 | 67 | |||
| AK209323.1 | - | 1, 2 | 67 | |||
| AK211228.1 | - | 1, 2 | 67 | |||
| AK214368.1 | - | 1, 2 | 67 | |||
| AK216934.1 | - | 1, 2 | 67 | |||
| AK218788.1 | - | 1, 2 | 67 | |||
| BC025487.1 | - | 2, 3 | 272 | |||
| BC141314.1 | - | 2, 3 | 309 | 9055926 | ||
| BC141315.1 | - | 2, 3 | 309 | 9055927 | ||

Cln5
Genomic Map