siRNA Details
| Gene Symbol: Pank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145962.2 | NP_666074.1 | 2 | 564 | ||
| GenBank mRNA | AK039865.1 | - | 2 | 566 | ||
| AK136305.1 | - | 2 | 254 | |||
| BC027089.1 | - | 2 | 176 | 5355443 | ||
| BC032188.1 | - | 2 | 610 | |||

Pank3
Genomic Map