siRNA Details
| Gene Symbol: 6720489N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173381.1 | NP_775557.1 | 3 | 1208 | ||
| GenBank mRNA | AK033001.1 | - | 3 | 1210 | ||
| AK033958.1 | - | 3 | 1192 | |||
| BC028252.1 | - | 1292 | ||||
| BC053725.1 | - | 3 | 1197 | |||

6720489N17Rik
Genomic Map