siRNA Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146180.1 | NP_001139652.1 | 10 | 1464 | ||
| NM_144800.2 | NP_659049.2 | 11 | 1476 | |||
| GenBank mRNA | AK030533.1 | - | 9 | 870 | ||
| AK129139.1 | - | 10 | 1461 | |||
| AY214918.1 | - | 11 | 977 | |||
| BC023682.1 | - | 5 | 582 | |||
| BC024131.1 | - | 11 | 1441 | 5101888 | ||

Mtss1
Genomic Map