siRNA Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144800.2 | NP_659049.2 | 14, 15 | 2082 | ||
| GenBank mRNA | AK030533.1 | - | 12, 13 | 1476 | ||
| AY214918.1 | - | 14, 15 | 1583 | |||
| BC016555.1 | - | 3, 4 | 397 | |||
| BC023682.1 | - | 8, 9 | 1188 | |||
| BC024131.1 | - | 14, 15 | 2047 | 5101888 | ||
| BC042632.1 | - | 3, 4 | 401 | |||

Mtss1
Genomic Map