siRNA Details
| Gene Symbol: Tmem87a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110496.1 | NP_001103966.1 | 7 | 618 | ||
| NM_001110497.1 | NP_001103967.1 | 7 | 618 | |||
| NM_173734.3 | NP_776095.2 | 7 | 618 | |||
| GenBank mRNA | AK044591.1 | - | 7 | 572 | ||
| AK086738.1 | - | 7 | 568 | |||
| AK088559.1 | - | 7 | 600 | |||
| AK134268.1 | - | 7 | 581 | |||
| AK137137.1 | - | 7 | 584 | |||
| BC004047.1 | - | 7 | 567 | |||
| BC027354.1 | - | 7 | 591 | 4010477 | ||

Tmem87a
Genomic Map