siRNA Details
| Gene Symbol: Tifa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145133.3 | NP_660115.1 | 3 | 557 | ||
| GenBank mRNA | AB062111.1 | - | 3 | 354 | ||
| AK041891.1 | - | 2 | 391 | |||
| AK044221.1 | - | 2 | 501 | |||
| AK149408.1 | - | 3 | 393 | |||
| AK149864.1 | - | 3 | 525 | |||
| AK151651.1 | - | 3 | 508 | |||
| AK166239.1 | - | 3 | 599 | |||
| BC065775.1 | - | 3 | 552 | 1528121 | ||

Tifa
Genomic Map