siRNA Details
| Gene Symbol: Mrgprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203490.3 | NP_987075.1 | 2 | 780 | ||
| GenBank mRNA | AY762266.1 | - | 2 | 780 | ||
| BC137948.1 | - | 2 | 686 | 8860967 | ||
| BC137950.1 | - | 2 | 686 | 8860969 | ||
More Information
Set Membership: |
|

Mrgprd
Genomic Map