siRNA Details
| Gene Symbol: Ptchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093750.1 | NP_001087219.1 | 3 | 2644 | ||
| GenBank mRNA | BC116312.1 | - | 3 | 2642 | 40095445 | |
| BC116313.1 | - | 3 | 2639 | 40095446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptchd1| Gene Symbol: Ptchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093750.1 | NP_001087219.1 | 3 | 2644 | ||
| GenBank mRNA | BC116312.1 | - | 3 | 2642 | 40095445 | |
| BC116313.1 | - | 3 | 2639 | 40095446 | ||