siRNA Details
| Gene Symbol: Fancd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033244.3 | NP_001028416.2 | 25 | 2426 | ||
| GenBank mRNA | AK142384.1 | - | 25 | 2424 | ||
| AK155602.1 | - | 25 | 2405 | |||
| AK166411.1 | - | 25 | 2396 | |||
| BC040776.1 | - | 13 | 1307 | |||
| BC042619.1 | - | 13 | 1307 | |||
| BC042870.1 | - | 1 | 36 | |||
| BC139004.1 | - | 25 | 2424 | 8862026 | ||

Fancd2
Genomic Map