siRNA Details
| Gene Symbol: Asap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004364.2 | NP_001004364.2 | 8 | 996 | ||
| NM_001098168.1 | NP_001091637.1 | 8 | 996 | |||
| NM_001135192.1 | NP_001128664.1 | 8 | 996 | |||
| GenBank mRNA | AK147622.1 | - | 8 | 1004 | ||
| AK147656.1 | - | 8 | 942 | |||
| AK162242.1 | - | 295 | ||||
| AK172944.1 | - | 8 | 808 | |||
| BC080847.1 | - | 8 | 996 | 6852788 | ||
| BC096022.1 | - | 8 | 764 | 30542525 | ||
| BM944240.1 | - | 3 | 191 | |||

Asap2
Genomic Map