siRNA Details
| Gene Symbol: Dennd6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134465.2 | NP_001127937.1 | 17, 18 | 1586 | ||
| NM_001285466.1 | NP_001272395.1 | 15, 16 | 1432 | |||
| NM_001285467.1 | NP_001272396.1 | 17, 18 | 1647 | |||
| NM_145969.4 | NP_666081.1 | 17, 18 | 1586 | |||
| GenBank mRNA | AK033036.1 | - | 15, 16 | 1434 | ||
| AK054197.1 | - | 17, 18 | 1530 | |||
| AK054295.1 | - | 17, 18 | 1587 | |||
| AK078910.1 | - | 1, 2 | 85 | |||
| AK080327.1 | - | 7, 8 | 527 | |||
| AK131163.1 | - | 17, 18 | 1558 | |||
| AK132219.1 | - | 17, 18 | 1583 | |||
| AK161908.1 | - | 17, 18 | 1556 | |||
| AK163396.1 | - | 17, 18 | 1646 | |||
| BC025603.1 | - | 17, 18 | 1581 | 5250476 | ||
| BC031553.1 | - | 3, 4 | 248 | |||
| BC050872.1 | - | 3, 4 | 238 | |||

Dennd6a
Genomic Map