siRNA Details
| Gene Symbol: Plekhh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181073.3 | NP_851418.2 | 8 | 1617 | ||
| GenBank mRNA | AK122464.1 | - | 8 | 1586 | ||
| AK143387.1 | - | 8 | 1434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhh1| Gene Symbol: Plekhh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181073.3 | NP_851418.2 | 8 | 1617 | ||
| GenBank mRNA | AK122464.1 | - | 8 | 1586 | ||
| AK143387.1 | - | 8 | 1434 | |||