siRNA Details
| Gene Symbol: Zfyve26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008550.1 | NP_001008550.1 | 24, 25 | 4929 | ||
| GenBank mRNA | AK220333.1 | - | 24, 25 | 4900 | ||
| BC047061.1 | - | 4, 5 | 495 | |||
| BC048379.1 | - | 4, 5 | 506 | |||
| BC141359.1 | - | 24, 25 | 4912 | 9055971 | ||

Zfyve26
Genomic Map