siRNA Details
| Gene Symbol: Zfyve26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008550.1 | NP_001008550.1 | 17 | 3185 | ||
| GenBank mRNA | AK041668.1 | - | 17 | 3182 | ||
| AK132344.1 | - | 17 | 3175 | |||
| AK220333.1 | - | 17 | 3156 | |||
| BC141359.1 | - | 17 | 3168 | 9055971 | ||

Zfyve26
Genomic Map