siRNA Details
| Gene Symbol: Tmem18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172049.2 | NP_742046.2 | 2, 3 | 244 | ||
| GenBank mRNA | AK133493.1 | - | 2, 3 | 244 | ||
| AK160817.1 | - | 2, 3 | 236 | |||
| AK166323.1 | - | 2, 3 | 261 | |||
| AK170496.1 | - | 2, 3 | 227 | |||
| BC024093.1 | - | 2, 3 | 216 | 5116657 | ||
| BY013255.1 | - | 2, 3 | 245 | |||

Tmem18
Genomic Map