siRNA Details
| Gene Symbol: Clrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178669.5 | NP_848784.1 | 3 | 684 | ||
| GenBank mRNA | AK033715.1 | - | 3 | 644 | ||
| AK085267.1 | - | 3 | 623 | |||
| AK146608.1 | - | 3 | 685 | |||
| BC036176.1 | - | 3 | 672 | 4989703 | ||

Clrn3
Genomic Map