siRNA Details
| Gene Symbol: Trim52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198601.3 | NP_941003.1 | 1 | 1063 | ||
| GenBank mRNA | AK029521.1 | - | 1 | 1069 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim52| Gene Symbol: Trim52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198601.3 | NP_941003.1 | 1 | 1063 | ||
| GenBank mRNA | AK029521.1 | - | 1 | 1069 | ||