siRNA Details
| Gene Symbol: Proser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173382.1 | NP_775558.2 | 8, 9 | 1099 | ||
| GenBank mRNA | AK034174.1 | - | 8, 9 | 1250 | ||
| AK038404.1 | - | 8, 9 | 1099 | |||
| AK087909.1 | - | 8, 9 | 1121 | |||
| BC086674.1 | - | 8, 9 | 656 | 30653602 | ||

Proser1
Genomic Map