siRNA Details
| Gene Symbol: 8030462N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178670.3 | NP_848785.2 | 2 | 971 | ||
| GenBank mRNA | AK033209.1 | - | 2 | 958 | ||
| AK046785.1 | - | 2 | 950 | |||
| AK139552.1 | - | 2 | 868 | |||
| AK163840.1 | - | 972 | ||||
| BC120889.2 | - | 1 | 681 | 40060928 | ||
| DQ465969.1 | - | 1 | 374 | |||
| DQ465970.1 | - | 1 | 122 | |||
| DQ465977.1 | - | 374 | ||||
| DQ465978.1 | - | 256 | ||||

8030462N17Rik
Genomic Map