siRNA Details
| Gene Symbol: Zswim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172503.3 | NP_766091.2 | 14 | 2880 | ||
| GenBank mRNA | AK052168.1 | - | 14 | 2894 | ||
| AK053663.1 | - | 14 | 2894 | |||
| AK131119.1 | - | 9 | 1738 | |||
| AK155465.1 | - | 14 | 2894 | |||
| BC016518.1 | - | 4 | 699 | |||
| BC058080.1 | - | 14 | 2880 | 6822376 | ||

Zswim4
Genomic Map