siRNA Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153535.2 | NP_705763.1 | 2 | 1206 | ||
| GenBank mRNA | AK034229.1 | - | 2 | 433 | ||
| AK038552.1 | - | 2 | 421 | |||
| AK140440.1 | - | 2 | 1206 | |||
| AK141607.1 | - | 2 | 1149 | |||
| AK198284.1 | - | 2 | 147 | |||
| AK212095.1 | - | 2 | 147 | |||
| AK220518.1 | - | 2 | 442 | |||
| BC035537.1 | - | 2 | 1097 | 5369152 | ||

Fam149a
Genomic Map