siRNA Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153535.2 | NP_705763.1 | 14 | 2837 | ||
| GenBank mRNA | AK034229.1 | - | 14 | 2064 | ||
| AK038552.1 | - | 14 | 2061 | |||
| AK140440.1 | - | 14 | 2837 | |||
| AK141607.1 | - | 14 | 2780 | |||
| AK187651.1 | - | 2 | 98 | |||
| AK220518.1 | - | 14 | 2082 | |||
| BC035537.1 | - | 14 | 2728 | 5369152 | ||

Fam149a
Genomic Map