siRNA Details
| Gene Symbol: Lcor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172154.4 | NP_742166.1 | 3 | 3944 | ||
| GenBank mRNA | AB076079.1 | - | 8 | 1555 | ||
| AK041090.1 | - | 3 | 3945 | |||
| AK041621.1 | - | 8 | 1758 | |||
| AK129442.1 | - | 6 | 1380 | |||
| AK155007.1 | - | 8 | 1634 | |||
| BC050068.1 | - | 3 | 926 | |||
| BC090652.1 | - | 5 | 1338 | 30657664 | ||
| BC099938.1 | - | 8 | 1626 | 30085146 | ||

Lcor
Genomic Map