siRNA Details
| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033246.2 | NP_001028418.1 | 6 | 2076 | ||
| GenBank mRNA | AK132742.1 | - | 6 | 2077 | ||
| AK161248.1 | - | 6 | 2064 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc110| Gene Symbol: Ccdc110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033246.2 | NP_001028418.1 | 6 | 2076 | ||
| GenBank mRNA | AK132742.1 | - | 6 | 2077 | ||
| AK161248.1 | - | 6 | 2064 | |||