siRNA Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153783.4 | NP_722478.2 | 4 | 1057 | ||
| GenBank mRNA | AF226656.2 | - | 4 | 1076 | ||
| AK010469.1 | - | 3 | 707 | |||
| AK030664.1 | - | 4 | 1058 | |||
| AK159137.1 | - | 2 | 344 | |||
| AK178754.1 | - | 2 | 247 | |||
| AK186235.1 | - | 2 | 211 | |||
| AM049402.1 | - | 4 | 1049 | |||
| BC033913.1 | - | 3 | 321 | |||
| BC082783.1 | - | 4 | 1056 | 6475809 | ||

Paox
Genomic Map