siRNA Details
| Gene Symbol: Sprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183147.2 | NP_898970.1 | 2 | 995 | ||
| GenBank mRNA | AK049995.1 | - | 2 | 1032 | ||
| BC056484.1 | - | 2 | 995 | 5696462 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprn| Gene Symbol: Sprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183147.2 | NP_898970.1 | 2 | 995 | ||
| GenBank mRNA | AK049995.1 | - | 2 | 1032 | ||
| BC056484.1 | - | 2 | 995 | 5696462 | ||