siRNA Details
| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145972.4 | NP_666084.1 | 5, 6 | 615 | ||
| GenBank mRNA | AK137274.1 | - | 4, 5 | 1308 | ||
| AK144612.1 | - | 5, 6 | 596 | |||
| AK162291.1 | - | 4, 5 | 457 | |||
| BC027231.1 | - | 5, 6 | 618 | 3585175 | ||

BC027231
Genomic Map