siRNA Details
| Gene Symbol: Aldh4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175438.4 | NP_780647.3 | 3 | 471 | ||
| GenBank mRNA | AK044712.1 | - | 3 | 267 | ||
| BC024133.1 | - | 2 | 189 | |||
| BC039281.1 | - | 3 | 255 | 4975491 | ||
| BC056226.1 | - | 3 | 355 | 5064715 | ||
| BC060650.1 | - | 3 | 471 | 30354228 | ||
| CA494657.1 | - | 3 | 508 | |||

Aldh4a1
Genomic Map