siRNA Details
| Gene Symbol: Satb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139146.2 | NP_631885.1 | 4 | 719 | ||
| GenBank mRNA | AF319623.1 | - | 4 | 719 | ||
| AJ606045.1 | - | 3 | 437 | |||
| AK129270.1 | - | 3 | 1710 | |||
| BC138625.1 | - | 3 | 470 | 8861645 | ||
| BC138626.1 | - | 3 | 470 | 8861646 | ||

Satb2
Genomic Map