siRNA Details
| Gene Symbol: Rsph4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162957.1 | NP_001156429.1 | 1 | 188 | ||
| GenBank mRNA | BC138000.1 | - | 1 | 188 | 8861019 | |
| BC138001.1 | - | 1 | 188 | 8861020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rsph4a| Gene Symbol: Rsph4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162957.1 | NP_001156429.1 | 1 | 188 | ||
| GenBank mRNA | BC138000.1 | - | 1 | 188 | 8861019 | |
| BC138001.1 | - | 1 | 188 | 8861020 | ||