siRNA Details
| Gene Symbol: Dse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172508.2 | NP_766096.1 | 6 | 2390 | ||
| GenBank mRNA | AK031737.1 | - | 6 | 2077 | ||
| AK045065.1 | - | 6 | 2297 | |||
| AK150737.1 | - | 6 | 2554 | |||
| AK153326.1 | - | 6 | 2063 | |||
| BC053095.1 | - | 6 | 2390 | 6401066 | ||

Dse
Genomic Map