siRNA Details
| Gene Symbol: Best2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130194.1 | NP_001123666.1 | 4 | 510 | ||
| GenBank mRNA | AY450428.1 | - | 4 | 752 | ||
| BC019528.1 | - | 3 | 484 | 4208860 | ||
| BC031186.1 | - | 3 | 471 | 4989959 | ||
| BC036157.1 | - | 4 | 510 | 4982413 | ||
| BC036163.1 | - | 4 | 551 | 4984410 | ||

Best2
Genomic Map