siRNA Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145218.3 | NP_660253.2 | 5 | 1769 | ||
| GenBank mRNA | AB175684.1 | - | 5 | 847 | ||
| AF467979.1 | - | 5 | 1050 | |||
| AK035817.1 | - | 5 | 1523 | |||
| AK048758.1 | - | 5 | 1514 | |||
| AK051281.1 | - | 5 | 2169 | |||
| AK087726.1 | - | 2339 | ||||
| BC052469.1 | - | 5 | 1165 | 6810287 | ||
| BC069177.1 | - | 5 | 1769 | |||
| BC158110.1 | - | 5 | 1796 | 9088184 | ||

Wbscr17
Genomic Map