siRNA Details
| Gene Symbol: Tnpo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122843.1 | NP_001116315.1 | 17 | 2130 | ||
| NM_145390.4 | NP_663365.3 | 17 | 2112 | |||
| GenBank mRNA | AK029774.1 | - | 7 | 1005 | ||
| AK142270.1 | - | 16 | 2667 | |||
| AK154985.1 | - | 17 | 2114 | |||
| AK155760.1 | - | 17 | 2114 | |||
| BC003275.1 | - | 17 | 2066 | 3499838 | ||

Tnpo2
Genomic Map