siRNA Details
| Gene Symbol: Ifitm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033632.1 | NP_001028804.1 | 1 | 123 | ||
| GenBank mRNA | AK039626.1 | - | 1 | 139 | ||
| BC104338.1 | - | 1 | 123 | 40049250 | ||
| BC104339.1 | - | 1 | 123 | 40049257 | ||

Ifitm6
Genomic Map