siRNA Details
| Gene Symbol: Gm4301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166637.1 | NP_001160109.1 | 1 | 67 | ||
| GenBank mRNA | BC147487.1 | - | 1 | 67 | 9056243 | |
| Gene Symbol: Gm4312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166636.1 | NP_001160108.1 | 1 | 67 | ||

Gm4301
Genomic Map